missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ SilberBrand™ ETERNA Type 2 Graduated Pipettes
AR-Glas™ graduated pipettes
Brand: BRAND™ 27002
This item is not returnable.
View return policy
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
To Deliver | |
B | |
12 | |
1 mL |
DIN EN ISO 835 | |
0.01 mL | |
Yellow | |
360 mm |