missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambion™ Silencer™ Select Custom Designed siRNA
Silencer™ custom siRNAs (unmodified 21-bp duplexes) are cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types.
£222.00 - £516.00
Specifications
Description | GeneAssist - Custom Designed Silencer™ Select siRNA |
---|---|
Product Line | Silencer™ Select |
Product Type | Custom Designed siRNA |
Shipping Condition | Ambient |
Temperature (Metric) | Upon receipt, store in a non-frost-free freezer at or below -20°C |
Product Code | Brand | Purity | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Purity | Quantity | Price | Quantity & Availability | ||||
15501054
|
Ambion™
4399665 |
Standard | 5 nmol |
£222.00
5nmol |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15511054
|
Ambion™
4399666 |
Standard | 20 nmol |
£330.00
20nmol |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15521054
|
Ambion™
4399667 |
Standard | 40 nmol |
£383.00
40nmol |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15531054
|
Ambion™
4399668 |
HPLC | 20 nmol |
£493.00
20nmol |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15541054
|
Ambion™
4399669 |
HPLC | 40 nmol |
£516.00
40nmol |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types
- Use the GeneAssist™ Custom siRNA Builder to design and order a custom siRNA
Specifications
GeneAssist - Custom Designed Silencer™ Select siRNA | |
Custom Designed siRNA | |
Upon receipt, store in a non-frost-free freezer at or below -20°C |
Silencer™ Select | |
Ambient |
For Research Use Only. Not for use in diagnostic procedures.