Learn More
Ambion™ Silencer™ Select Pre-Designed siRNA
Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry.
Brand: Ambion™ 4427037
98.80 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
- Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry
- Silencer™ Select Pre-Designed siRNAs are guaranteed-to-silence based on their proven design
- Sets of Silencer™ Select siRNA Libraries are also available in tubes or plate for high throughput screening
- The Silencer™ Select Positive and Negative Control RNAs should be included in every siRNA experiment.
The benefits of using Silencer™ Select siRNAs include:
- Potent – up to 100-fold more potent than currently available siRNAs and fewer off-target effects
- Most Specific – LNA™ chemical modifications reduce off-target effects by up to 90%
- Reliable – demonstrated improvements in consistency and reliability of phenotypic results
- Guaranteed – 100% guaranteed to silence, the best guarantee in the industry
Note: Silencer™ Pre-Designed & Validated siRNAs are designed using our first-generation algorithm and DO NOT include any chemical modifications. They are recommended for completing in vitro RNAi studies that were started using Silencer™ siRNAs. For new studies we recommend Silencer Select siRNAs.
Approximate Turnaround Times*
- Standard purification: 4 business days
- HPLC purification: 6 business days (20 and 40 nmol); 15 business days (250 nmol)
- In Vivo Ready: 15 business days (250 nmol)
Specifications
Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below -20°C. | |
siRNA | |
Silencer™ Select | |
1 nmol |
Silencer™ Select Pre-Designed siRNA | |
RNAi, epigenetics and non-coding RNA research, RNAi, siRNA | |
Standard | |
Store at -20°C |
For Research Use Only. Not for use in diagnostic procedures.