missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambion™ Silencer™ Select Pre-Designed siRNA
Silencer™ Select Pre-Designed siRNA
Brand: Ambion™ 4390818
335.00 GBP valid until 2024-09-27
Use promo code "21650" to get your promotional price.
This item is not returnable.
View return policy
Description
The Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry. A choice of Silencer™ Select siRNAs are available—Pre-Designed or Validated. Silencer™ Select Pre-Designed siRNAs are guaranteed-to-silence based on their proven design while the Silencer™ Select Validated siRNAs are functionally tested to reduce target gene expression. Sets of Silencer™ Select siRNA Libraries are also available in tubes or plate for high throughput screening. The Silencer™ Select Positive and Negative Control RNAs should be included in every siRNA experiment.The benefits of using Silencer™ Select siRNAs include:
Note: Silencer™ Pre-Designed & Validated siRNAs are designed using our first-generation algorithm and DO NOT include any chemical modifications. They are recommended for completing in vitro RNAi studies that were started using Silencer™ siRNAs. For new studies we recommend Silencer Select siRNAs.
Approximate Turnaround Times*
Specifications
Contains dried siRNA (40 nmol) and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °. | |
siRNA | |
Silencer™ Select pre-designed siRNA | |
40 nmol |
Silencer™ Select Pre-designed siRNA, 40nmol, HPLC Purity, –20°C Storage, Potent, Most Specific, Reliable, Guaranteed, Incorporate the Latest Improvements in siRNA design, Off-Target Effect Prediction Algorithms and Chemistry | |
Silencer™ Select, Ambion™ | |
HPLC |
For Research Use Only. Not for use in diagnostic procedures.