missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambion™ Silencer™ Select Pre-Designed siRNA
Silencer™ Select Pre-Designed siRNA
£242.00 - £366.00
Specifications
RNAi Type | siRNA |
---|---|
Final Product Type | Silencer™ Select pre-designed siRNA |
For Use With (Application) | RNAi, epigenetics and non-coding RNA research, RNAi, siRNA |
Product Line | Silencer™ Select, Ambion™ |
Product Type | Silencer™ Select pre-designed siRNA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10147724
|
Ambion™
4390771 |
5 nmol |
£242.00
Each |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10157724
|
Ambion™
4390815 |
20 nmol |
£318.00
Each |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10704617
|
Ambion™
4390816 |
40 nmol |
£366.00
Each |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry. A choice of Silencer™ Select siRNAs are available—Pre-Designed or Validated. Silencer™ Select Pre-Designed siRNAs are guaranteed-to-silence based on their proven design while the Silencer™ Select Validated siRNAs are functionally tested to reduce target gene expression. Sets of Silencer™ Select siRNA Libraries are also available in tubes or plate for high throughput screening. The Silencer™ Select Positive and Negative Control RNAs should be included in every siRNA experiment.The benefits of using Silencer™ Select siRNAs include:
Note: Silencer™ Pre-Designed & Validated siRNAs are designed using our first-generation algorithm and DO NOT include any chemical modifications. They are recommended for completing in vitro RNAi studies that were started using Silencer™ siRNAs. For new studies we recommend Silencer Select siRNAs.
Approximate Turnaround Times*
Specifications
siRNA | |
RNAi, epigenetics and non-coding RNA research, RNAi, siRNA | |
Silencer™ Select pre-designed siRNA |
Silencer™ Select pre-designed siRNA | |
Silencer™ Select, Ambion™ | |
Standard purity |
For Research Use Only. Not for use in diagnostic procedures.