Learn More
Ambion™ Silencer™ Validated siRNA
Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry.
Brand: Ambion™ AM51342
1490.07 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
- The Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry
- Silencer™ Select Validated siRNAs are functionally tested to reduce target gene expression
- Sets of Silencer™ Select siRNA Libraries are also available in tubes or plate for high throughput screening
- The Silencer™ Select Positive and Negative Control RNAs should be included in every siRNA experiment
The benefits of using Silencer™ Select siRNAs include:
- Potent – up to 100-fold more potent than currently available siRNAs and fewer off-target effects
- Most Specific – LNA™ chemical modifications reduce off-target effects by up to 90%
- Reliable – demonstrated improvements in consistency and reliability of phenotypic results
- Guaranteed – 100% guaranteed to silence, the best guarantee in the industry
Note: Silencer™ Pre-Designed & Validated siRNAs are designed using our first-generation algorithm and DO NOT include any chemical modifications. They are recommended for completing in vitro RNAi studies that were started using Silencer™ siRNAs. For new studies we recommend Silencer Select siRNAs.
Approximate Turnaround Times*
- Standard purification: 4 business days
- HPLC purification: 6 business days (20 and 40 nmol); 15 business days (250 nmol)
- In Vivo Ready: 15 business days (250 nmol)
Specifications
Store at room temperature | |
siRNA | |
In Vivo | |
Store at room temperature |
Silencer™ Validated siRNA | |
Silencer™ | |
1 μmol |
For Research Use Only. Not for use in diagnostic procedures.