missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Silencer™ Select Validated siRNA

Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry.

Brand:  Ambion™ 4444247

5910.90 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15591084

  • £10370.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Découvrez plus d'offres spéciales
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

  • The Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry
  • Silencer™ Select Validated siRNAs are functionally tested to reduce target gene expression
  • Sets of Silencer™ Select siRNA Libraries are also available in tubes or plate for high throughput screening
  • The Silencer™ Select Positive and Negative Control RNAs should be included in every siRNA experiment

The benefits of using Silencer™ Select siRNAs include:

  • Potent – up to 100-fold more potent than currently available siRNAs and fewer off-target effects
  • Most Specific – LNA™ chemical modifications reduce off-target effects by up to 90%
  • Reliable – demonstrated improvements in consistency and reliability of phenotypic results
  • Guaranteed – 100% guaranteed to silence, the best guarantee in the industry

Note: Silencer™ Pre-Designed & Validated siRNAs are designed using our first-generation algorithm and DO NOT include any chemical modifications. They are recommended for completing in vitro RNAi studies that were started using Silencer™ siRNAs. For new studies we recommend Silencer Select siRNAs.

Approximate Turnaround Times*

  • Standard purification: 4 business days
  • HPLC purification: 6 business days (20 and 40 nmol); 15 business days (250 nmol)
  • In Vivo Ready: 15 business days (250 nmol)
TRUSTED_SUSTAINABILITY
Spécification

Spécification

Store in freezer (-5° to -30°C).
siRNA
HPLC
Store in freezer (-5° to -30°C)
Silencer™ Select Validated siRNA
Silencer™ Select
10 μmol
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.