Learn More
Silencer™ siRNA Library, In Plate
Silencer™ siRNA Library, In Plate
Brand:  Invitrogen AM81810
Product Code. 10288794
  • £16.59 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Silencer™ siRNA Libraries feature combinations of Silencer™ Pre-designed siRNAs, and when available, Silencer™ Validated siRNAs, in 96-well or 384-well plates.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Room temperature
Library SiRNA, 1μmol, Powder, Room Temperature Storage, In Plate, Custom, feature Combinations of Silencer Pre-designed siRNAs and When Available, Silencer Validated siRNAs, in 96-well or 384-well Plates
Library SiRNA
1 μmol