Learn More
Thermo Scientific™ SilTite™ Replacement Ferrules , Nuts and Baseplate Seals
Product Code. 10659275 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Agilent SSL Injectors
All SilTite Kits
MS Interface
Thermo Scientific DSQ GC-MS Interface
Thermo Scientific DSQ GC-MS Interface Kits
Thermo Scientific TRACE 1300/1310 SSL and ISQ GC-MS Interface Kits
Includes:
Pk. of 10 Ferrules
Pk. of 5 Nuts
Description:
290 MA201
290 MA202
290 MA203
290 MA205
290 MA207
290 MT211
290 MT221
290 MT222
290 MT223
Unit Size:
Pack of 10
Pack of 5
This item is not returnable. Consulta la politica di reso
Codice prodotto. 10659275
missing translation for 'mfr': Thermo Scientific™ 290MA203
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule.
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule, which forms a strong, permanent, airtight seal around the capillary column.
  • SilTite metal ferrules and nuts are made from the same material and have the same thermal
  • expansion coefficient
  • Base of the ferrule is flat and forms a perfect seal with the MS interface
  • The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
  • Unlike other ferrules, do not need re-tightening after installation
TRUSTED_SUSTAINABILITY
Specifica
Type Replacement Ferrule
Description 290 MA203
For Use With (Equipment) All SilTite Kits
Material SilTite Metal
Includes Pk. of 10 Ferrules
Diameter (Metric) Inside 0.53 mm
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato