missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica 100-200μm, 60Å, EcoChrom™, MP Biomedicals™
For Chromatography; Chemical Synthesis
£42.82 - £1335.60
Specifications
Content And Storage | Store at Room Temperature. |
---|---|
Type | Silica |
Physical Form | Powder |
MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
Density | 2g/cm3 at 20°C |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
17867034
|
MP Biomedicals
0202760 |
500g |
£42.82
500g |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11383549
|
MP Biomedicals
0202761 |
25kg |
£1335.60
25kg |
Estimated Shipment: 26-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at Room Temperature. | |
Powder | |
2g/cm3 at 20°C |
Silica | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
7631-86-9 |