missing translation for 'onlineSavingsMsg'
Learn More

Silica 63-100μm, 60Å, EcoChrom™, MP Biomedicals™

For Chromatography; Chemical Synthesis

£1810.00

Specifications

Content And Storage Store at Room Temperature.
Type Silica
Physical Form Powder
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Density 2g/cm3 at 20°C
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11383599
View Documents
MP Biomedicals
0204660
25 kg
£1810.00
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Store at Room Temperature.
Powder
2g/cm3 at 20°C
Silica
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
7631-86-9
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silica 63-100μm, 60Å, EcoChrom™, MP Biomedicals™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.