Learn More
Silica Gel (2.5-6.0mm) Self Indicating, Dessicant Agent, Fisher Chemical
Brand: Fisher Chemical S/0762/53
40.75 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
500g, Plastic powder jar

Specifications
White | |
2.0 to 3.5 | |
500 g | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08g/mol |
Solid | |
7631-86-9 | |
O2Si | |
Plastic Powder Jar | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Fisher Chemical Silica gel, 4 to 7 mesh (2.5mm to 6.0mm), self indicating
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
14Hours
11Minutes
26Seconds
8
Your input is important to us. Please complete this form to provide feedback related to the content on this product.