Learn More
Silica Gel 60 Angstrom Pore Size, 35-70 Micron, for Column Chromatography, Fisher Chemical™
Brand: Fisher Chemical S/0693/65
482.34 GBP valid until 2024-07-31
Use promo code "21824" to get your promotional price.
Code : 25
Additional Details : CAS Number : 7631-86-9 Weight : 5.00000kg
Description
Silica Gel 60 Angstrom Pore Size, 35-70 Micron, for Column Chromatography
Specifications
Plastic Keg | |
5 kg |