missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica gel, C18-RP, 17%C, ca.0,8mmol/g, part. size: 40-63μ, Thermo Scientific Chemicals
£157.00 - £628.13
Chemical Identifiers
CAS | 71889-02-6 |
---|---|
Molecular Formula | C18H37Cl3O2Si2 |
Molecular Weight (g/mol) | 448.012 |
InChI Key | VYTRPUAMVCWPLO-UHFFFAOYSA-N |
Synonym | Octadecylsilicate |
PubChem CID | 172932 |
IUPAC Name | dioxosilane;trichloro(octadecyl)silane |
SMILES | CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl.O=[Si]=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10111002
|
Thermo Scientific Acros
360120500 |
50 g | Glass Bottle |
£157.00
50g |
Estimated Shipment: 05-12-2024
Log in to see stock. |
||||
11924121
|
Thermo Scientific Acros
360122500 |
250 g | Glass Bottle |
£628.13
250g |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatographyChemical Identifiers
71889-02-6 | |
448.012 | |
Octadecylsilicate | |
dioxosilane;trichloro(octadecyl)silane |
C18H37Cl3O2Si2 | |
VYTRPUAMVCWPLO-UHFFFAOYSA-N | |
172932 | |
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl.O=[Si]=O |
Specifications
71889-02-6 | |
Octadecylsilicate | |
VYTRPUAMVCWPLO-UHFFFAOYSA-N | |
dioxosilane;trichloro(octadecyl)silane | |
172932 |
C18H37Cl3O2Si2 | |
Solubility in water: insoluble. | |
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl.O=[Si]=O | |
448.012 | |
Silica gel, C18-RP, 17%C |
Safety and Handling
Product Identifier
- Silicagel, C18-Reversed Phase
RUO – Research Use Only