missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, C8-RP, 12%C, ca. 1.2 mmol/g, part. size 40-63 micron

Product Code. p-4112268 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic Bottle
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 10540352

Brand: Thermo Scientific Acros 360140500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00148339
Synonym Octylsilicate

Specifications

Color White
Physical Form Powder
Material Silica Gel
Quantity 50 g
Packaging Plastic Bottle
Particle Size 40 to 63 Micron
Chemical Name or Material Silica gel, C8-RP, 12%C, ca. 1.2 mmol/g, part. size 40-63 micron

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.