missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Click to view available options
Quantity:
1 kg
5 kg
30 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
Specifications
Specifications
Pore Size | 60 Å |
Quantity | 30 kg |
Type | Silica Gel |
Material | Silica Gel |
For Use With (Application) | Column Chromatography |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'