missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, for chromatography, 0.035-0.070 mm, 90 A, Thermo Scientific Chemicals

£26.33 - £78.09

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym Silicon dioxide
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10721804
View Documents Promotion Details
Thermo Scientific Acros
241652500
250 g Plastic Bottle
£26.33
250g
Please to purchase this item. Need a web account? Register with us today!
10732661
View Documents Promotion Details
Thermo Scientific Acros
241650010
1 kg Plastic Bottle
£78.09
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography
Specifications

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Silicon dioxide
CHEBI:30563
O=[Si]=O

Specifications

7631-86-9
O2Si
Silicon dioxide
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.08
Silica Gel
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Solubility in water: insoluble.
O=[Si]=O
60.08
CHEBI:30563
Silica gel, For chromatography, 0.035-0.070mm, 90Å
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Silica gel

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only