missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Silica gel, for column chromatography, 0.035-0.070mm, 60Å Product Code.: 10273542

Thermo Scientific Chemicals Silica gel, for column chromatography, 0.035-0.070mm, 60Å

Product Code. 10273542
5 kg
Click to view available options
Quantity:
1 kg
5 kg
30 kg
This item is not returnable. View return policy

Product Code. 10273542

Brand: Thermo Scientific Chemicals 240360050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9

Specifications

Pore Size 60 Å
Packaging Plastic Drum
Quantity 5 kg
Type Silica Gel
Material Silica Gel
For Use With (Application) Column Chromatography

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466636 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number240360050,240360300,240360010
Lot NumberA0459321 Certificate TypeCertificate of Analysis Date15/01/2024 Catalog Number240360050,240360300,240360010
Lot NumberA0452114 Certificate TypeCertificate of Analysis Date26/04/2023 Catalog Number240360050,240360300,240360010
Lot NumberA0391957 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number240360050,240360300,240360010
Lot NumberA0395911 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number240360050,240360300,240360010
Product Identifier
  • Silica gel

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Silica gel, for column chromatography, 0.035-0.070mm, 60Å >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.