missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Silica gel, for column chromatography, 0.060-0.200mm, 40Å Product Code.: 11904121

Thermo Scientific Chemicals Silica gel, for column chromatography, 0.060-0.200mm, 40Å

Product Code. 11904121
5 kg
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11904121

Brand: Thermo Scientific Chemicals 241660050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9

Specifications

Pore Size 40 Å
Packaging Plastic Drum
Quantity 5 kg
Type Silica Gel
Material Silica Gel
Molecular Formula O2Si
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name silanedione
Molecular Weight (g/mol) 60.08
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465711 Certificate TypeCertificate of Analysis Date04/09/2024 Catalog Number241660050,241660010
Lot NumberA0462435 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number241660050,241660010
Lot NumberA0457565 Certificate TypeCertificate of Analysis Date17/01/2024 Catalog Number241660050,241660010
Lot NumberA0401258 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number241660050,241660010
Lot NumberA0418251 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number241660050,241660010
Product Identifier
  • Silica gel

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Silica gel, for column chromatography, 0.060-0.200mm, 40Å >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.