missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Silica gel, for column chromatography, Ult Product Code.: 10689852

Thermo Scientific Chemicals Silica gel, for column chromatography, Ult

Product Code. 10689852
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 10689852

Brand: Thermo Scientific Chemicals 360031000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9

Specifications

Particle Size 40 to 60 μm
Pore Size 40 Å
Packaging Plastic Bottle
Quantity 100 g
Type Silica Gel
Material Silica Gel
For Use With (Application) Column Chromatography

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0435301 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number360031000,360035000
Lot NumberA0431076 Certificate TypeCertificate of Analysis Date01/01/2024 Catalog Number360031000,360035000
Lot NumberA0396702 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number360031000,360035000
Lot NumberA0415276 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number360031000,360035000
Lot NumberA0409825 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number360031000,360035000
Product Identifier
  • Silica gel

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Silica gel, for column chromatography, Ult >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.