missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, for drying purposes, non-toxic grade, 3-6 mm

Product Code. p-41554 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Plastic Bottle
Plastic Drum
Unit Size:
1kg
25kg
5kg
This item is not returnable. View return policy

Product Code. 10440983

Brand: Thermo Scientific Acros 357400010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Silica gel, O2Si, CAS Number-7631-86-9, Silicon dioxide, 25kg, 2.0% max. (140 deg.C, 4 hrs), Adsorption capacity (relative humidity 40%): ca 20%, 620 to 800g/L, CHEBI:30563, White to Yellow, 231-545-4, 60.08, Non-toxic, VYPSYNLAJGMNEJ-UHFFFAOYSA-N, dioxosilane, 15, 8632, MFCD00011232 | CAS: 7631-86-9 | O2Si | 60.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent, For column chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym Silicon dioxide
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Color White to Yellow
Physical Form Solid
Density 620 to 800g/L
Material Silica Gel
Quantity 1 kg
Moisture Residual moisture (140°C): < 1% (typically)
Merck Index 15, 8632
Packaging Plastic Bottle
Particle Size Particle size analysis: typically sieve residue>5mm:ca 10%, passing through 2mm: ca 1%
Solubility Information Solubility in water: insoluble.
IUPAC Name silanedione
Formula Weight 60.08
Grade Non-toxic
Water Absorption 20.0% min. (40% min. RH)
Chemical Name or Material Silica gel, 3-6mm
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.