missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel Rubin Drying Bags, Honeywell Fluka™

With indicator function, 1/3 TE, bag with ∼10 g, crystalline

Brand:  Honeywell Fluka 80148-1KG

Code : NEW

Additional Details : CAS Number : 112926-00-8 Weight : 1.00000kg

 View more versions of this product

Product Code. 15657160

  • £144.39 / 1kg
Estimated Shipment: 16-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Silica gel Rubin drying bags
Silica Gel
O2Si
Metal Can
O=[Si]=O
60.08
112926-00-8
1 kg
NONH for all modes of transport
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers