Learn More
Silica Gel Rubin Drying Bags, Honeywell Fluka™
With indicator function, 1/3 TE, bag with ∼10 g, crystalline
Brand: Honeywell Fluka 80148-250G
Code : NEW
Additional Details : CAS Number : 112926-00-8 Weight : 0.25000kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Silica gel Rubin drying bags | |
Silica Gel | |
O2Si | |
Metal Can | |
O=[Si]=O | |
60.08 |
112926-00-8 | |
250 g | |
NONH for all modes of transport | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
silanedione |