missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel Rubin Drying Bags, Honeywell Fluka™

With indicator function, 1/3 TE, bag with ∼10 g, crystalline

Brand:  Honeywell Fluka 72811-1KG

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15686910

  • £176.39 / 1kg
Estimated Shipment: 01-10-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

1600°C
Red
Silica gel Rubin drying bags
2g/cm3
1 kg
Poly Bottle
2230°C
9
112926-00-8
Silica Gel
NONH for all modes of transport
Slightly soluble in water
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers