missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica Gel Rubin, Honeywell Fluka™
62.37 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 2230°C |
Color | Red |
Melting Point | 1600°C |
Physical Form | Granular |
pH | 9.0 |
Material | Silica Gel |
Quantity | 1 kg |
UN Number | NONH for all modes of transport |
Odor | Odorless |
Packaging | Poly Bottle |
Show More |
Safety and Handling
Product Identifier
- Silica gel Rubin
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'