missing translation for 'onlineSavingsMsg'
Learn More
Silica gel, TLC high purity grade, without binder, Thermo Scientific Chemicals Product Code.: 11335557

Silica gel, TLC high purity grade, without binder, Thermo Scientific Chemicals

Product Code. 11335557
50 g
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11335557

Brand: Thermo Scientific Alfa Aesar 042756.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For TLC Plate Preparation

Silica gel, TLC high purity grade, without binder is employed for thin layer chromatography plate preparation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
MDL Number MFCD00011232
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
Synonym metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Color White
Physical Form Powder
Quantity 50 g
Solubility Information Soluble in alkalies. Insoluble in water.
Grade TLC High Purity
Odor Odorless
Chemical Name or Material Silica gel

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silica gel, TLC high purity grade, without binder, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.