Learn More
Macherey-Nagel™ Silica N/UV254 Adsorbents
Brand:  Macherey-Nagel™ 816340.1
 View more versions of this product
Product Code. 10587041
  • £74.10 / 1kg
Estimated Shipment: 06-11-2024
to see stock.
Explore more special offers
Add to basket
Specifications
1 kg
2 to 20 μm
0.75 mL/g
Silica N/UV254 Adsorbent
Silica
60 Å