missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Non-Deactivated Fused Silica GC Tubing

Used in a wide range of capillary GC, LC and Bioanalytical applications. Trajan Scientific and Medical™ Non-Deactivated Fused Silica GC Tubing is protected with a high temperature Polyimide resin

Brand:  Trajan™ 062482

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15218843

  • £241.00 / 25m
Estimated Shipment: 14-08-2024
to see stock.
Add to basket

Description

Description

  • Used in a wide range of capillary GC, LC and Bioanalytical applications
  • Made from high quality fused silica
  • Tubing protected with a high temperature Polyimide resin
  • 0.005mm-0.53mm internal diameters
Specifications

Specifications

Capillary Tubing
GC, LC and Bioanalytical Applications
0.68 mm
Silica
Not deactivated
0.53 mm
25 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions