missing translation for 'onlineSavingsMsg'
Learn More

Silicagel orange, for drying purposes, non toxic grade, Thermo Scientific Chemicals

Product Code. 10005313
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic Drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1327-36-2
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.04
MDL Number MFCD00077645
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N

Specifications

Color Orange
Physical Form Beads
Material Silica Gel
Quantity 250 g
Loss on Drying 2% max. (120°C, 2 h)
Packaging Glass Bottle
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Silica gel orange

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.