missing translation for 'onlineSavingsMsg'
Learn More
Silicic Acid, 99.9%, Honeywell™ Fluka™ Product Code.: 15622940

Silicic Acid, 99.9%, Honeywell™ Fluka™

Product Code. 15622940 Shop All Honeywell Fluka Products
25 g, Poly Bottle
Click to view available options
Quantity:
25 g
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15622940

Brand: Honeywell-Fluka 28877225G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purified by refining, 20 μm, 99.9%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

pH 3 to 8 (at 33g/L 25°C)
CAS 1343-98-2
Packaging Poly Bottle
Quantity 25 g
Molecular Formula O2Si
MDL Number MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
UN Number NONH for all modes of transport
Synonym orthosilicic acid, monosilicic acid, tetrahydroxysilane, h4sio4, silicon tetrahydroxide, silicasol, kieselsaeure, si oh 4, unii-623b93yabh, orthosilicicacid
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name silicic acid
Molecular Weight (g/mol) 60.08
PubChem CID 14942
ChEBI CHEBI:26675
Percent Purity 99.90%
Chemical Name or Material Silicic Acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicic Acid, 99.9%, Honeywell™ Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.