missing translation for 'onlineSavingsMsg'
Learn More
Silicic Acid, n-Hydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™ Product Code.: 15527814

Silicic Acid, n-Hydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15527814
500 g, Poly Bottle
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15527814

Brand: Avantor J.T.Baker 032401

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
Synonym metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Packaging Poly Bottle
Specific Gravity 1L = 2.10kg
Quantity 500 g
Physical Form Powder
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Silicic Acid, n-Hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicic Acid, n-Hydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.