missing translation for 'onlineSavingsMsg'
Learn More
Silicon carbide, beta-phase, nanopowder, Thermo Scientific Chemicals Product Code.: 11445480

Silicon carbide, beta-phase, nanopowder, Thermo Scientific Chemicals

Product Code. 11445480
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11445480

Brand: Thermo Scientific Alfa Aesar 044646.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

S.A. 70 to 90 m2/g

Silicon carbide can be used form very hard ceramics that are widely used in applications requiring high endurance, such as car brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide as light-emitting diodes (LEDs) and detectors in early radios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicideShow MoreShow Less
PubChem CID 9863
ChEBI CHEBI:29390
SMILES [C-]#[Si+]

Specifications

Melting Point 2700°C
Color Black to Gray
Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water
IUPAC Name methanidylidynesilylium
Formula Weight 40.1
Odor Odorless
Chemical Name or Material Silicon carbide, beta-phase

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU09K024 Certificate TypeCertificate of Analysis Date14/08/2024 Catalog Number044646.22,044646.14,044646.06
Lot NumberZ14I020 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog Number044646.22,044646.14,044646.06
Lot NumberU06G036 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044646.22,044646.14,044646.06
Lot NumberP24I044 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044646.22,044646.14,044646.06
Lot NumberW20H017 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044646.22,044646.14,044646.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicon carbide, beta-phase, nanopowder, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.