missing translation for 'onlineSavingsMsg'
Learn More

Silicon carbide powder, fine, 320 grit, Thermo Scientific Chemicals

£47.41 - £430.14

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
PubChem CID 9863
ChEBI CHEBI:29390
IUPAC Name methanidylidynesilanylium
SMILES [C-]#[Si+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11437947
SDS View Documents
Thermo Scientific Alfa Aesar
A16601.36
500 g
£47.41
500g
Please to purchase this item. Need a web account? Register with us today!
11447947
SDS View Documents
Thermo Scientific Alfa Aesar
A16601.0E
2500 g
£175.88
2500g
Please to purchase this item. Need a web account? Register with us today!
11457947
SDS View Documents
Thermo Scientific Alfa Aesar
A16601.0C
10,000 g
£430.14
10000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Silicon carbide is used in abrasives, in polishing and grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic. It compositely plays an important role in the crack-healing behavior.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon carbide is used in abrasives, in polishing and grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic. It compositely plays an important role in the crack-healing behavior.

Solubility
Soluble in molten sodium hydroxide, potassium hydroxide and in molten iron.

Notes
Keep it away from strong oxidizing agents.
Specifications

Chemical Identifiers

409-21-2
40.10
HBMJWWWQQXIZIP-UHFFFAOYSA-N
9863
methanidylidynesilanylium
CSi
MFCD00049531
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
CHEBI:29390
[C-]#[Si+]

Specifications

409-21-2
MFCD00049531
Soluble in molten sodium hydroxide,potassium hydroxide and in molten iron.
[C-]#[Si+]
40.10
CHEBI:29390
3.16g/mL
CSi
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
HBMJWWWQQXIZIP-UHFFFAOYSA-N
methanidylidynesilanylium
9863
40.1
Silicon carbide, fine, 320 grit
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.

Warning

missing translation for 'einecsNumber' : 206-991-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.