missing translation for 'onlineSavingsMsg'
Learn More

Silicon carbide whiskers, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11339037
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Abrasive For Polishing/ Grinding

Used in abrasives, polishing, grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are the light-emitting diodes and sensors. It is utilitarian for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic, compositely play an important role in crack-healing behavior.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
PubChem CID 9863
ChEBI CHEBI:29390
IUPAC Name methanidylidynesilanylium
SMILES [C-]#[Si+]

Specifications

Melting Point 2700°C
Color Black to Gray
Quantity 10 g
Assay Percent Range 99% (metals basis)
Solubility Information Soluble in molten alkalis (NaOH,KOH) and molten iron. Insoluble in water.
Formula Weight 40.1
Odor Odorless
Density 3.2
Chemical Name or Material Silicon carbide whiskers

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'