missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon Dioxide, calcined, crude, Honeywell Fluka™
Click to view available options
Quantity:
2.5 kg
50 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 6 x 2.5 kg |
Linear Formula | SiO2 |
UN Number | NONH for all modes of transport |
Formula Weight | 60.08 |
Chemical Name or Material | Silicon Dioxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'