missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon diOxide, purum p.a., Acid purified, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
5 kg
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
Quantity | 500 g |
Linear Formula | SiO2 |
UN Number | NONH for all modes of transport |
Formula Weight | 60.08g/mol |
Density | 2.6g/mL at 25°C |
Chemical Name or Material | Silicon dioxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'