Learn More
Saint Gobain Life Sciences Silicon Folding Skirt Stopper
Product Code. p-8004866
Click to view available options
Diameter (Metric):
7.1 mm
16 mm
Unit Size:
Pack of 100
Pack of 20
Product Code. 11744598
Brand: Saint Gobain Life Sciences 467007100
Only null left
Add to basket
Add to basket
Use this stopper for biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus. Silicon Folding Skirt Stopper is particularly suited for all types of tissue culture work.
  • Two-part folding skirt stoppers
  • Designed with a double seal to keep container contents secure
  • Ideal for securing glass, metal and plastic containers
  • The lower section grips the outside of the container and the upper hollow plug features annular serrations to provide a tight seal at the rim
  • Upper hollow plug section can be perforated to extract contents without removing the stopper and remains watertight even after several perforations
  • Biologically neutral and non-toxic silicone caps for closing tubes and flasks
  • Odorless, tasteless, non-adherent and resistant to aging
  • Nontoxic, biologically neutral
  • FDA/BGA approved
  • For test tubes
  • Easy to install
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 7.1 mm
Quantity 1 Ea.
Color Transparent
Material Silicone
Product Type Folding Skirt Stopper