missing translation for 'onlineSavingsMsg'
Learn More

Silicon granular, virgin poly fines, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 15456255
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.09
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]

Specifications

Quantity 25 g
Percent Purity 99.999%
Odor Odorless
Particle Size ∽1/4 in. & down
Form Chunks to Powder
Assay (metals basis)
Chemical Name or Material Silicon granular, Virgin poly fines

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'