missing translation for 'onlineSavingsMsg'
Learn More
Silicon(II) oxide, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11328858

Silicon(II) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11328858
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11328858

Brand: Thermo Scientific Alfa Aesar 041711.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For vacuum deposition

Silicon(II) oxide can be used for synthesis of fine ceramic materials, such as silicon nitride, silicon carbide fine ceramic powder raw materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10097-28-6
Molecular Formula OSi
Molecular Weight (g/mol) 44.084
MDL Number MFCD00151536
InChI Key LIVNPJMFVYWSIS-UHFFFAOYSA-N
Synonym silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilyleneShow MoreShow Less
PubChem CID 66241
ChEBI CHEBI:30588
IUPAC Name oxoniumylidynesilanide
SMILES [O+]#[Si-]

Specifications

Boiling Point 1880°C
Melting Point >1702°C
Physical Form Pieces
Quantity 100 g
Merck Index 14,8496
Solubility Information Insoluble in water.
Formula Weight 44.09
Percent Purity 99.9%
Odor Odorless
Density 2.13 g/mL
Assay (metals basis)
Chemical Name or Material Silicon(II) oxide, 6 to 15mm Pieces
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW14J013 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog Number041711.22
Lot NumberX10F104 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number041711.22
Lot NumberJ31L37 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041711.22
Lot NumberG23S014 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041711.22
Lot NumberU01D008 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041711.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicon(II) oxide, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.