Learn More
Silicon(IV) acetate, Thermo Scientific Chemicals
Description
Silicon tetraacetate is used in the preparation of silicon dioxide thin films by a direct photochemical vapor deposition method. It serves as a precursor to prepare silicon complexes with monofunctional bidentate Schiff bases. It is also used as an alternative to silicon hydride and alkoxide for low-temperature silicon dioxide production. Further, it reacts with ethanol in the absence of water to get silica gel and ethyl acetate. In addition, it is employed as a sol-gel precursor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 160°C to 170°C (decomposition) |
Melting Point | 110°C |
Physical Form | Crystalline |
Quantity | 250 g |
UN Number | UN3261 |
Merck Index | 14,8498 |
Solubility Information | Soluble in acetone and benzene. |
Formula Weight | 264.27 |
Sensitivity | Moisture sensitive |
Chemical Name or Material | Silicon(IV) acetate |
RUO – Research Use Only