missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) bromide, Thermo Scientific Chemicals

Product Code. 11391559
Click to view available options
Quantity:
10 g
50 g

Silicon(IV) bromide is used to grow thin films of silicon by the chemical vapor deposition method. It is also used for ceramics, sealants and the production of many cutting tools.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-66-4
Molecular Formula Br4Si
Molecular Weight (g/mol) 347.701
MDL Number MFCD00049530
InChI Key AIFMYMZGQVTROK-UHFFFAOYSA-N
Synonym silicon iv bromide, silicon tetrabromide, silane, tetrabromo, sibr4, unii-8gac9ywl42, 8gac9ywl42, silicon bromide, acmc-20ajqj
PubChem CID 82247
IUPAC Name tetrabromosilane
SMILES [Si](Br)(Br)(Br)Br

Specifications

Boiling Point 154°C
Melting Point 5°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 50 g
UN Number UN3264
Merck Index 14,8499
Formula Weight 347.72
Odor Stench
Sensitivity Moisture sensitive
Density 2.800 g/mL
Chemical Name or Material Silicon(IV) bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'