missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) iodide, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11329638
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Silicon(IV) iodide is used as a precursor to prepare silicon amides. It finds application in the manufacture and etching of silicon in microelectronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-84-4
Molecular Formula I4Si
Molecular Weight (g/mol) 535.70
MDL Number MFCD00054041
InChI Key CFTHARXEQHJSEH-UHFFFAOYSA-N
Synonym silicon tetraiodide, silane, tetraiodo, silicon iv iodide, periodosilane, tetraiodosilicon, acmc-1bu37, silicon tetraiodide trace metals basis, silicon iv iodide trace metals basis 1g
PubChem CID 83498
IUPAC Name tetraiodosilane
SMILES I[Si](I)(I)I

Specifications

Boiling Point 290°C
Melting Point 120°C
Color White
Physical Form Lump
Quantity 5 g
Linear Formula SiI4
UN Number UN2923
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 535.7
Percent Purity 99.999%
Sensitivity Air, light, and moisture sensitive
Density 4.198 g/mL
Assay (metals basis)
Chemical Name or Material Silicon (IV) iodide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'