missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, 15% in H{2}O, colloidal dispersion

Product Code. 11324187
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11324187

Brand: Thermo Scientific Alfa Aesar 036669.A7

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-Temperature Binders, Anti-Slip Coatings For Paper, Petrochemical Catalyst Support and Binder | CAS: 7631-86-9 | O2Si | 60.08 g/mol

High-temperature binders, anti-slip coatings for paper, petrochemical catalyst support and binder, polishing agent, abrasion resistant coatingsSilicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of food and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 650 m2/g
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Physical Form Particles in Liquid
Quantity 5 kg
Solubility Information Immiscible with water.
IUPAC Name silanedione
Formula Weight 60.09
Percent Purity 15%
Chemical Name or Material Silicon(IV) oxide, 15% in H2O

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.