missing translation for 'onlineSavingsMsg'
Learn More
Silicon(IV) oxide, microcrystalline, Thermo Scientific Chemicals Product Code.: 11352118

Silicon(IV) oxide, microcrystalline, Thermo Scientific Chemicals

Product Code. 11352118
2 kg
Click to view available options
Quantity:
2 kg
10 kg
This item is not returnable. View return policy

Product Code. 11352118

Brand: Thermo Scientific Alfa Aesar 013024.A3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

In the manufacture of glass, ceramics and abrasives, in water filtration, in cosmetics, pharmaceuticals and rubber, as a thermal insulatorSilica is used in the manufacture of glass for windows, drinking glasses, beverage bottles, and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware, and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Melting Point 1710°C
Color Brown to White
Physical Form Powder
Quantity 2 kg
Assay Percent Range 99.5%
Solubility Information Insoluble in water and acids. Soluble in HF. Very slightly soluble in alkalies
IUPAC Name dioxosilane
Formula Weight 60.09
Odor Odorless
Chemical Name or Material Silicon(IV) oxide, 98.5-99% microcrystalline silicon in the form of quartz
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN06L007 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog Number013024.A9,013024.A3
Lot NumberZ13D013 Certificate TypeCertificate of Analysis Date24/07/2023 Catalog Number013024.A9,013024.A3
Lot NumberZ20I037 Certificate TypeCertificate of Analysis Date19/01/2023 Catalog Number013024.A9,013024.A3
Lot NumberE31Z041 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number013024.A9,013024.A3
Lot NumberK19S017 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number013024.A9,013024.A3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicon(IV) oxide, microcrystalline, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.