missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon(IV) oxide, Amorphous fumed, S.A. 175 to 225m2/g, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
Used as sodium stabilizing counterion, for preparation of silica catalytic supports. Also used in glass manufacturing, foundry sand casting, metallurgical flux sand, ceramics sand, gas recovery, paint and coatings, filtration sand, chromatographic column treatment, wood treatment, inks, desiccants and powder metal molding. Its is principle gateway in electronic industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
S.A. 175 to 225m2/g
Specifications
Specifications
Boiling Point | 2230°C |
Melting Point | 1710°C |
Color | White |
Physical Form | Powder |
Quantity | 50 g |
Solubility Information | Insoluble in water. |
IUPAC Name | dioxosilane |
Formula Weight | 60.09 |
Odor | Odorless |
Density | 2.2 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'