Learn More
Silicon(IV) oxide, amorphous fumed, S.A. 350-420m2/g, Thermo Scientific Chemicals
Description
Functionalized RAFT agent for controlled radical polymerization; especially suited for the polymerization of styrene; acrylate and acrylamide monomers. Azide group can be used to conjugate to a variety of alkyne-functionalized biomolecules. Chain Transfer Agent (CTA). SDS mixture of sodium alkyl sulfates consisting chiefly of sodium lauryl sulfate. Used as a suspending and thickening agent in pharmaceutical
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 2230°C |
Melting Point | 1710°C |
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Merck Index | 14,8493 |
Solubility Information | Insoluble in water. |
Formula Weight | 60.09 |
Odor | Odorless |
Refractive Index | 1.46 |
Show More |
RUO – Research Use Only