missing translation for 'onlineSavingsMsg'
Learn More
Silicon(IV) oxide, Amorphous fumed, S.A. 85 to 115m2/g, Thermo Scientific Chemicals Product Code.: 11385527

Silicon(IV) oxide, Amorphous fumed, S.A. 85 to 115m2/g, Thermo Scientific Chemicals

Product Code. 11385527
1 kg
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11385527

Brand: Thermo Scientific Alfa Aesar 042737.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For Adhesives, Sealants, Silicones

Silicon(IV) oxide, amorphous is used as carriers, processing aids, anti-caking and free-flow agents in animal feed. Defoamer applications such as paint, food, paper, textile and other industrial applications. Synthetic silicon dioxides are used as a rheology control agent in plastics. It is also used to manufacture adhesives, sealants and silicones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 85 to 115m2/g
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Melting Point 1710°C
Color White
Physical Form Powder
Quantity 1 kg
Solubility Information Insoluble in water.
Formula Weight 60.09
Odor Odorless
Refractive Index 1.46
Density 2.2
Chemical Name or Material Silicon(IV) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberI20W038 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog Number042737.30,042737.18,042737.A1
Lot NumberE02T007 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number042737.30,042737.18,042737.A1
Lot NumberE12K16 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number042737.30,042737.18,042737.A1
Lot NumberK24T049 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number042737.30,042737.18,042737.A1
Lot NumberK07N37 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number042737.30,042737.18,042737.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicon(IV) oxide, Amorphous fumed, S.A. 85 to 115m2/g, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.