missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, powder, 0.5 micron, 99.9%, Thermo Scientific Chemicals

Silicon(IV) oxide, 99.9%, O2Si, CAS Number-7631-86-9, silica, amorphous, silicic anhydride, tridymite, infusorial earth, silicon dioxide, silica, cristobalite, quartz, aerosil, diatomaceous earth, 100g, 2230 deg.C, CHEBI:30563, 231-545-4, 60.09, Warning

£64.32 - £206.03

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11363876
View Documents
Thermo Scientific Alfa Aesar
L16985.14
25 g
£64.32
25g
Please to purchase this item. Need a web account? Register with us today!
11373876
View Documents
Thermo Scientific Alfa Aesar
L16985.22
100 g
£206.03
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

Solubility
Insoluble in water.

Notes
It is a very stable compound of silicon.
Specifications

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
O=[Si]=O

Specifications

2230°C
7631-86-9
O2Si
14,8493
Insoluble in water.
O=[Si]=O
60.08
CHEBI:30563
99.9%
2
1710°C
Powder
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.09
Odorless
Silicon(IV) oxide, 0.5 micron
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 231-545-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only