Learn More
Silicon Powder, crystalline, -100+200 mesh, 99.99% (Metals basis), Thermo Scientific Chemicals
£32.55 - £223.11
Chemical Identifiers
CAS | 7440-21-3 |
---|---|
Molecular Formula | Si |
Molecular Weight (g/mol) | 28.09 |
MDL Number | MFCD00085311 |
InChI Key | XUIMIQQOPSSXEZ-UHFFFAOYSA-N |
Synonym | metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 |
PubChem CID | 5461123 |
ChEBI | CHEBI:27573 |
IUPAC Name | silicon |
SMILES | [Si] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15467015
|
Thermo Scientific Alfa Aesar
045609.09 |
10 g | N/A | ||||||
15477015
|
Thermo Scientific Alfa Aesar
045609.18 |
50 g |
£86.53
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15487015
|
Thermo Scientific Alfa Aesar
045609.30 |
250 g |
£223.11
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-21-3 | |
28.09 | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
5461123 | |
silicon |
Si | |
MFCD00085311 | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831Show More | |
CHEBI:27573 | |
[Si] |
Specifications
7440-21-3 | |
Crystalline Powder | |
Si | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
[Si] | |
28.09 | |
CHEBI:27573 | |
Gray to black powder |
Black to Gray | |
99.99% (Metals basis) | |
MFCD00085311 | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
silicon | |
5461123 | |
Odorless | |
Silicon |
Safety and Handling
H228-H303: Flammable solid. May be harmful if swallowed.
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III
missing translation for 'einecsNumber' : 231-130-8
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.