Learn More
Fisherbrand™ Silicon Platinum-cured Tubing
Ideal to use with peristaltic pump due to its higher tear strength and less vulnerability towards splitting or mechanical damage.
£21.55 - £248.00
Specifications
Certifications/Compliance FDA, BGA and WRC approved
View More Specs
Products 17
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
11532563
View Documents
Fisherbrand™
FB50853
3 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11542573
View Documents Promotion Details
Fisherbrand™
FB50875
14 mm
£248.00
10m
In Stock
to see stock.
Edge
Add to basket
11552573
View Documents
Fisherbrand™
FB50877
17 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11562573
View Documents
Fisherbrand™
FB50879
22.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11582573
View Documents
Fisherbrand™
PCST/25X31.5
31.5 mm
£106.61
5m
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
11572573
View Documents
Fisherbrand™
PCST/20X26.5
26.5 mm
£90.90
5m
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
11552563
View Documents
Fisherbrand™
FB50857
4.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11562563
View Documents
Fisherbrand™
FB50859
5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11572563
View Documents
Fisherbrand™
FB50861
6 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11582563
View Documents
Fisherbrand™
FB50863
7.2 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11592563
View Documents
Fisherbrand™
FB50865
8 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11502573
View Documents
Fisherbrand™
FB50867
9 mm
£112.32
10m
In Stock
to see stock.
Edge
Add to basket
11512573
View Documents
Fisherbrand™
FB50869
9.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11522573
View Documents
Fisherbrand™
PCST/8X12
12 mm
£170.00
10m
In Stock
to see stock.
Edge
Add to basket
11532573
View Documents
Fisherbrand™
FB50873
13 mm
£173.00
10m
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
12951388
View Documents
Fisherbrand™
PCST/8X14.4
14.4 mm
£49.50
5m
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
11542563
View Documents
Fisherbrand™
PCST/2X4
4 mm
£37.40
10m
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Tube has no smell, taint or reaction with chlorine
  • Platinum cured tubing has higher clarity with no yellowing, as experienced with peroxide cured materials
  • 60°IRHD ±5°C
  • FDA, BGA and WRC approved
  • Temperature range: -60°C to 200°C
  • Anti-adherent, hydrophobic
  • High electrical and ozone resistance
  • Less bacteriological build up
  • Can be used for food, laboratory, pharmaceutical and medical applications
Specifications
Specifications
FDA, BGA and WRC approved
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Silicon Platinum-cured Tubing